Learn More
uvex Phynomic All-Round Safety Gloves
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
5
6
7
8
9
10
11
12
Dimensione della confezione:
10 paia
Descrizione
- Lightweight construction acts as a second-skin to maintain a high level of feeling
- Aqua-polymer foam coating is moisture repellent and flexible
- Breathable materials help prevent excessive perspiration
- Dirt-resistant exterior is ideal for mechanical tasks
- Enhanced grip in dry and slightly damp applications
- 3D Ergo technology ensures a secure fit
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
- Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
Specifica
Specifica
| Certificazione/Conformità | OEKO-TEX™ Standard 100 |
| Applicazioni | Industrial |
| Coating Material | Aqua-Polymer Foam |
| Colore | Gray, Black |
| Finitura esterna | Smooth |
| Hand | Ambidestri |
| Latex Free | Sì |
| Materiale | Polyamide, Elastane |
| Product Type | Mechanical Protection Gloves |
| Size | 6 |
| Vedi altri risultati |