Learn More
uvex Phynomic FOAM Protective Gloves with Aqua-Polymer Foam Coating
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
5
8
9
10
12
Dimensione della confezione:
10 paia
Pezzo
Descrizione
- High dexterity safety glove designed for precision mechanical work
- Damp-resistant, aqua-polymer foam coating provides mechanical abrasion resistance
- Highly-breathable palm coating enhances grip capabilities in dry and slightly damp areas
- Tactile feel when assembling small parts
- Ideal for precision assembly work, inspection, sorting, food processing, and packaging
Specifica
Specifica
| Struttura | Seamless Knit |
| Coating Material | Aqua-Polymer Foam |
| Colore | White, Gray |
| Finitura esterna | Smooth |
| Hand | Ambidestri |
| Materiale | Spandex, Polyamide |
| Product Type | Mechanical Protection Gloves |
| Size | 8 |
| Stile | Full-hand Mitt |
| EN388 Standard | 3121X |
| Vedi altri risultati |