Learn More
uvex Phynomic FOAM Protective Gloves with Aqua-Polymer Foam Coating
Codice prodotto. p-7236913 Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
5
8
9
10
12
Dimensione della confezione:
10 paia
Pezzo
Codice prodotto. 17757579
Marca: uvex 6005005
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Phynomic series gloves are ideal for all types of work where a high level of feeling is required. These safety gloves act as a second skin and are especially light and flexible, without making hands immediately sweaty. The aqua-polymer foam coating enhances grip capabilities in dry environments.
  • High dexterity safety glove designed for precision mechanical work
  • Damp-resistant, aqua-polymer foam coating provides mechanical abrasion resistance
  • Highly-breathable palm coating enhances grip capabilities in dry and slightly damp areas
  • Tactile feel when assembling small parts
  • Ideal for precision assembly work, inspection, sorting, food processing, and packaging
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità EN 388 (3 1 2 1 X)
Struttura Seamless Knit
Coating Material Aqua-Polymer Foam
Colore White, Gray
Finitura esterna Smooth
Hand Ambidestri
Materiale Spandex, Polyamide
Product Type Mechanical Protection Gloves
Size 5
Stile Full-hand Mitt
EN388 Standard 3121X
Stile polso guanto Knit
Serie Phynomic FOAM