Learn More
Thermo Scientific™ Sodium Acetate (3 M)
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
1 L
Dimensione della confezione:
1L
Descrizione
Thermo Scientific™ Pierce Sodium Acetate Solution is a sterile, 3M soluion of sodium acetate that is useful for DNA purification and storage.
Additional Information and Online Ordering
Thermo Scientific™ Pierce™ Sodium Acetate Solution is a sterile, 3M solution of sodium acetate that is useful for DNA purification and storage. The clear, colorless and odorless liquid is delivered in 1L bottles.
Specifica
Specifica
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. |
| Product Type | Sodium Acetate Solution |
| Quantità | 1 L |
| Forma | Liquid |