missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3C2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA587065
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Purity is > 95% by SDS-PAGE.
The Class II PI3 kinases, including PIK3C2A, phosphorylate the 3 position of PI or PI4P. Though not as well studied as the Class I PI3 kinases, the Class II enyzmes have been linked to diverse, receptor-mediated cellular processes including insulin signaling, neuronal survival, and growth factor signaling. PI3-kinases play roles in signaling pathways involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. This protein contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases. C2 domains act as calcium-dependent phospholipid binding motifs that mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The PI3-kinase activity of this protein is not sensitive to nanomolar levels of the inhibitor wortmanin. This protein was shown to be able to be activated by insulin and may be involved in integrin-dependent signaling.
Specifica
| PIK3C2A | |
| Polyclonal | |
| Unconjugated | |
| PIK3C2A | |
| C2-containing phosphatidylinositol kinase; CPK; Cpk-m; DKFZp686L193; MGC142218; p170; phosphatidylinositol 3-kinase, C2 domain containing, alpha polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha; phosphoinositide 3-kinase-C2-alpha; phosphoinositide-3-kinase, class 2, alpha polypeptide; PI3KC2; PI3-K-C2(ALPHA); pi3kc2a; PI3-K-C2A; PI3K-C2alpha; PI3K-C2-alpha; Pik3c2a; ptdIns-3-kinase C2 subunit alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18704, 361632, 5286 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00443, Q61194 | |
| PIK3C2A | |
| Recombinant protein of human PIK3C2A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto