missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP4K2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17077-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifica
| PIP4K2C | |
| Polyclonal | |
| Unconjugated | |
| PIP4K2C | |
| PI(5)P 4 kinase type II gamma, PIP4K2C, PIP4KII gamma, PIP5K2C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117150, 140607, 79837 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88370, Q8TBX8, Q91XU3 | |
| PIP4K2C | |
| PIP4K2C Fusion Protein Ag10989 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto