Learn More
PKC Gamma Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 29208-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC gamma is a neuron-specific member of the classical PKCs and is activated and translocated to subcellular regions as a result of various stimuli, including diacylglycerol synthesis, increased intracellular Ca(2+) and phorbol esters. Defects in this protein have been associated with spinocerebellar ataxia type 14 (SCA14), an autosomal dominant neurodegenerative disease.
Specifica
PKC Gamma | |
Polyclonal | |
Unconjugated | |
150 μL | |
5582 | |
Liquid |
Western Blot, ELISA | |
1000 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat |