missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP1-77266CL1
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PKR Polyclonal antibody specifically detects PKR in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifica
| PKR | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PKR. The immunogen is located within the last 50 amino acids of PKR. Amino Acid Squence: TVWKKSPEKNERHT | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Signal Transduction | |
| 5610 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto