Learn More
Cole-Parmer™ Plas-Labs™ 800-PRV Glove Box Pressure Relief Valve
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Overall Length (English):
1 in.
Dimensione della confezione:
Pezzo
Descrizione
- Acrylic relief valve.
- Features 1/8in. NPT stainless steel inlet and outlet ports.
- Exhausted air can be directed away from operators by a hose connection to the outlet port.
Specifica
Specifica
| Descrizione | NPT(M) x NPT(F) |
| Materiale | Stainless Steel |
| Overall Length (English) | 1 in. |
| Ingresso | 1/8 in. NPT(M) |
| Porta di uscita | 1/8 in. NPT(F) |
| Larghezza (sistema imperiale) | 1 in. |
| Altezza (sistema imperiale) | 2 in. |
| Product Type | Glove Box Pressure Relief Valve, 1/8 in. NPT Stainless Steel |