Learn More
DWK Life Sciences Plastic Hose Connection, with silicone seal (VMQ), PP complete with screw cap PBT, bent
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Quantità:
100/Pk.
Dimensione della confezione:
Pezzo
Descrizione
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PP are used for lower thermal and chemical resistance requirements.
- Hose olives enable the connection of hoses to laboratory apparatuses
Specifica
Specifica
| Quantità | 100/Pk. |
| Descrizione articolo | For applications with lower thermal and chemical resistance requirements |
| Joint Size | 8.6 mm |