Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Click to view available options
Capacità:
10 Tubes up to 30 mm
14 tubes up to 30mm
22 Tubes up to 15 mm
26 Tubes up to 15 mm, 1.5 -15 mL
48 Tubes up to 15 mm
5 x 30 mm and 12 x 15 mm tubes
8 x 30 mm, 22 x 15 mm tubes
Dimensione della confezione:
Pezzo
Descrizione
- For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
- Hold as few as 8 test tubes or as many as 48.
- Accommodates a variety of capacities
Specifica
Specifica
| Capacità | 14 tubes up to 30mm |