Learn More
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A
Codice prodotto. 30053181 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/8 in.
Diametro (sistema imperiale) esterno:
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
7/16 in.
Dimensione della confezione:
15m
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diametro (sistema imperiale) interno Diametro (sistema imperiale) esterno unitSize
30053181 1/4 in. 7/16 in. 15m
30053184 1/8 in. 1/4 in. 15m
30054183 1/4 in. 3/8 in. 15m
30053005 1/16 in. 3/16 in. 15m
30053899 3/8 in. 5/8 in. 15m
30054065 1/4 in. 1/2 in. 15m
30053651 3/16 in. 7/16 in. 15m
30053154 1/2 in. 3/4 in. 15m
30053638 3/16 in. 5/16 in. 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Codice prodotto. 30053181 Fornitore Cole-Parmer N. del fornitore 5011962
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
E in., hibits very good resilience, kink resistance, and pressure resistance
Superior optical clarity—easily view your sample. Odorless, noncytoto in., ic, and nonhemolytic. Low-binding surface helps minimize particle and odor absorption. Does not contain BPA, phthalates, PVC, or pero in., ides. Cleanroom packed and double bagged package. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifica
Certificazione/Conformità ADCF;FDA;Non-Phthalate;USP;RoHS
Diametro (sistema imperiale) interno 1/4 in.
Diametro (sistema imperiale) esterno 7/16 in.
Overall Length (English) 50 ft.