Learn More
Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)
Codice prodotto. 11314767
Click to view available options
Quantità:
25 x 25 mm
50 x 50 mm
Dimensione della confezione:
25mm
Pezzo
Codice prodotto. 11314767
Marca: Thermo Scientific Alfa Aesar 041807.FF
Richiesta di formato massivo o personalizzato
Pt:Ir; 80:20 wt%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Platinum Iridium foil
Quantità 25 x 25 mm
Formula molecolare Pt:Ir; 80:20 wt%
Numero MDL MFCD02091683
Percent Purity 99.9%
Imballaggio Tube