Learn More
Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)
Product Code. 11370899
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370899 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11370899 Supplier Thermo Scientific Alfa Aesar Supplier No. 010672.14
 Restricted Item
Spedizione stimata: 15-12-2026
Add to basket
CAS: 7440-06-4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 6-4-7440
Quantity 25 g
Percent Purity 99.99%