missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLP1 Monoclonal Antibody (plpc 1)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA190652
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLP1 is a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.This gene encodes a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.
Specifica
| PLP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P04116, P60201 | |
| PLP1 | |
| Synthetic peptide GRGTKF corresponding to C terminal region of myelin proteolipid protein. | |
| 50 μg | |
| Primary | |
| Human, Bovine, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| plpc 1 | |
| Unconjugated | |
| PLP1 | |
| DM20; GPM6C; HLD1; jimpy; jp; lipophilin; lipophilin (aa 129 to 276); major myelin proteolipid protein; MMPL; msd; Myelin proteolipid protein; myelin synthesis deficiency; PLP; PLP/DM20; Plp1; PMD; proteolipid; proteolipid protein; proteolipid protein (myelin) 1; Proteolipid protein (Pelizaeus-Merzbacher disease spastic paraplegia 2 uncomplicated); Proteolipid protein (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein 1; proteolipid protein 1 (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein, lipophilin; rsh; rump shaker; SPG2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 281410, 5354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto