Learn More
SI Analytics™ Combinazione di cavi con spina
Codice prodotto. 10001072 Sfoglia Tutto SI Analytics Prodotti
Cambia vista
Click to view available options
Lunghezza (sistema metrico) cavo:
1 m
2 m
5 m
10 m
Tipo:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 2 A
Plug Cable Combination
Dimensione della confezione:
Pezzo
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Lunghezza (sistema metrico) cavo Tipo unitSize
10001072 1 m L 1 NN Pezzo
10535553 5 m K 10 A Pezzo
10326932 1 m L 1 N Pezzo
10604671 1 m L 1 BNC Pezzo
10406812 1 m K 1 A Pezzo
10684661 1 m L 1 A Pezzo
10466165 2 m L 2 A Pezzo
10652832 5 m Plug Cable Combination Pezzo
10152071 2 m K2X Pezzo
10559371 10 m K5A Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Codice prodotto. 10001072 Fornitore SI Analytics™ N. del fornitore 285122489
Please to purchase this item. Need a web account? Register with us today!
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifica
Tipi di connettore 2 x 4 mm Banana Plug
Tipo L 1 NN
Include 1m Coaxial Cable, 2 x 4mm Banana Plug
Lunghezza (sistema metrico) cavo 1 m