Learn More
Memmert™ Prolunghe a innesto per forno
Use these tube extensions with Memmert ovens. Memmert™ Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.
Marca: Memmert™ B29719
Dettagli aggiuntivi : Peso : 0.79200kg
Descrizione
- Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
- Angled
- Only models U, I, S
Specifica
Exhaust Air Ducting | |
Prolunga tubo plug-in | |
2.2 (Inside) in., 2.4 (Outside) in. | |
57 (Inside) mm, 60.3 (Outside) mm |