Learn More
Abnova™ PLXNA2 (Human) Recombinant Protein
Human PLXNA2 full-length ORF ( AAH32125, 1 a.a. - 164 a.a.) recombinant protein with GST-tag at N-terminal.
Marca:  Abnova™ H00005362-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16129231
  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Sequence: MHSQVFSAFPYSLPLRFWVNVIKNPQFVFDIHKGSITDACLSVVAQTFMDSCSTSEHRLDKDSPSNKLLYAKDIPSYKSWVERYYADIAKLPAISDQDMNAYLAEQSRLHAVEFNMLSALNEIYSYVSKYSEELIGALEQDEQARRQRLAYKVEQLINAMSIES
Specifica
Specifica
AAH32125
plexin A2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PLXNA2
GST
5362
Wheat germ expression system
25 μg
FLJ11751, FLJ30634, KIAA0463, OCT, PLXN2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer