Learn More
PNRC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 51052-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Nuclear receptor coactivator. May play a role in signal transduction.Specifica
| PNRC1 | |
| Polyclonal | |
| Unconjugated | |
| PNRC1 | |
| B4 2, PNAS 145, PNRC1, PROL2, Proline rich protein 2, Protein B4 2, PRR2, RP11 63L7.5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10957 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q12796 | |
| PNRC1 | |
| PNRC1 Fusion Protein Ag0100 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |