Learn More
Podoplanin Mouse anti-Human, Unconjugated, Clone: 1D9F3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67432-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Podoplanin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Podoplanin. This antibody recognizes human antigen. The Podoplanin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifica
| Podoplanin | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunohistochemistry, ELISA | |
| 1D9F3 | |
| Unconjugated | |
| 150 μL | |
| 10630 | |
| IgG2a |