Learn More
Abnova™ POLR2H (Human) Recombinant Protein
Human POLR2H full-length ORF ( AAH00739, 1 a.a. - 150 a.a.) recombinant protein with GST-tag at N-terminal.
Marca:  Abnova™ H00005437-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16129321
  • 528.00€ / 25µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Sequence: MAGILFEDIFDVKDIDPEGKKFDRVSRLHCESESFKMDLILDVNIQIYPVDLGDKFRLVIASTLYEDGTLDDGEYNPTDDRPSRADQFEYVMYGKVYRIEGDETSTEAATRLSAYVSYGGLLMRLQGDANNLHGFEVDSRVYLLMKKLAF
Specifica
Specifica
AAH00739
polymerase (RNA) II (DNA directed) polypeptide H
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
POLR2H
GST
5437
Wheat germ expression system
25 μg
RPABC3, RPB17, RPB8, hsRPB8
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer