Learn More
MP Biomedicals™ Poly-L-Lysine Hydrochloride
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
100 mg
500 mg
Dimensione della confezione:
100mg
500mg
Descrizione
- Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates.
- Poly-L-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
- When adsorbed to the culture surface, poly-L-lysine increases the number of positively charged sites available for cell binding.
- Polymers of both D- and L-lysine are used to coat solid surfaces.
- Poly-L-lysine has been reported to improve the protein coating of ELISA plates.
Specifica
Specifica
| CAS | 26124-78-7 |
| Colore | Whitish |
| Content And Storage | -20°C |
| Formato | Powder |
| Da utilizzare con (applicazione) | Cellular attachment factor in culture applications |
| Peso molecolare | >20,000 g/mol |
| Quantità | 100 mg |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.