Learn More
Ekastu Safety™ Occhiali di protezione in policarbonato
Click to view available options
:
Colorless
Descrizione
- DIN EN 166 1 – FT
- Ideal for factory visits, assembly line and laboratory workers
- High mechanical stability, besides a robustly lightweight
- Made of transparent plastic, scratch-proof and anti-fog
- Excellent all-round view and safety
- Provides modern styling in combination with eyebrow and side protection
- 99.99% UV protection with innovative UV 380 technology
- Also suitable for wearer of glasses as over-spectacle
Specifica
Specifica
| Certificazione/Conformità | DIN EN 166 1 - FT |
| Lens Coating | Scratch-resistant |
| Descrizione | Plastica |
| Lens Tint | Filtro UV |
| Product Type | Occhiali di sicurezza |
| Colore telaio | Trasparente |
| N. per confezione | 1 ciasc. |
| Lens Material | Policarbonato |
| Materiale del telaio | Polycarbonate |
| UV Protection | UV Filter |