Learn More
Brady™ Etichette in poliestere stampabili con trasferimento termico
Click to view available options
Forma:
Rectangle
Rectangle with Rounded Corners
Rettangolo
Round
Dimensioni (sistema metrico):
101.6W x 50.8mmH
15.24 mm dia.
15.24 x 48.26 mm (W x H)
15.24 x 66.68 mm (W x H)
18.26W x 25.46 mmH
19 x 6.35 mm (W x H)
19W x 38.1 mmH
19W x 6.35 mmH
22.86W x 19 mmH
22.86W x 22.86 mmH
25.4 x 12.70 mm (W x H)
25.4W x 12.7mmH
25.4W x 4.75mmH
25.4W x 76.2mmH
25.4W x 9.53 mmH
31.75W x 19 mmH
34.93W x 45.72 mmH
38.10W x 6.35mmH
38.1W x 19 mmH
40W x 15 mmH
41.28 x 15.24 mm (W xH)
48.26W x 6.35 mmH
5.08W x 22.86 mmH
50.8W x 25.4 mmH
50.8W x 6.35 mmH
57.15W x 19 mmH
76.20W x 25.4mmH
76.20W x 50.8 mmH
76.2W x 25.4mmH
88.9W x 38.1 mmH
9.53 mm dia.
H 25.4 x L 50.8 mm
L 15.24 x H 38.1 mm
Finitura:
Gloss
Lucido
Opaco
Satin
Tipo di adesivo:
Acrilico permanente, Permanent
Acrylic, Permanent
Rubber, Permanent
Dimensione della confezione:
1 ROLL
1 confezione
1 rotolo
Descrizione
- Glossy white finish with permanent acrylic is thermal transfer printable
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Service temperature of -94 to 230° F (-70 to 110° C)
- Compatible printers: BBP11, BBP12, THT printer, 25 mm core
Specifica
Specifica
| Certificazione/Conformità | RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted |
| Descrizione | Thermal Transfer Printable Labels |
| Tipo di adesivo | Acrylic, Permanent |
| Rivestimento | None |
| Colore | White |
| Finitura | Gloss |
| Formato | Roll |
| Materiale | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Vedi altri risultati |