Learn More
Cole-Parmer™ Cella di polietilene dotata di guanti interni
Click to view available options
Lunghezza (sistema metrico):
431.8 mm
508 mm
685.8 mm
939.8 mm
Larghezza (sistema metrico):
431.8 mm
508 mm
635 mm
685.8 mm
939.8 mm
Altezza (sistema metrico):
279.4 mm
304.8 mm
381 mm
457.2 mm
939.8 mm
Dimensione della confezione:
pezzi 6
Descrizione
An alternative to standard glove boxes—these durable, polyethylene glove chambers are reusable, yet priced to be disposable. Chambers are easy to use—just slide your equipment through the sleeves, seal, connect the gas port (accepts 3⁄8'' ID tubing) to your gas supply, and inflate.
- Disposable chambers simplify cleanup
- Two equipment sleeves and two rear-mounted gas ports for added versatility
- Glove bags have a 2.5 mil thickness and are made of polyethylene
- Glove bags are not autoclaveable, nor are they sterile
- One pair of cotton gloves, tubing coupler and a closure included
Specifica
Specifica
| Materiale | Polietilene |
| Altezza (sistema metrico) | 381 mm |
| Include | One Pair Of Cotton Gloves, Tubing Coupler, and a Closure |
| Lunghezza (sistema metrico) | 685.8 mm |
| Quantità | 6/Pk. |
| Larghezza (sistema metrico) | 685.8 mm |