Learn More
Kartell™ Bottiglie graduate in polietilene con collo largo
Designed with a wide mouth for easy filling and emptying of liquid or powder samples. Kartell™ Graduated Wide-Mouth Bottles are for general purpose use.
Marca: Kartell™ 0161200
2.14 EUR valido fino al 2025-12-31
MIGLIOR PREZZO in promozione! Usa il codice promo "24145" per avere il tuo prezzo promozionale.
Descrizione
- Graduated general purpose bottles
- Long neck thread ensures leakproof closure
- Conforms to DIN 13316 and 168
- Suitable for foodstuff