Learn More
Kappes Systeme™ Scatola di trasporto Euro in polipropilene
Marca: Kappes Systeme™ 6476.00.4454
Dettagli aggiuntivi : Peso : 0.10000kg
Specifica
Transport Box | |
Gray | |
Conveyor, Grabber Technology, Storage and Retrieval Machine | |
1/Pk. | |
600 mm | |
420 mm |
Polypropylene | |
Rectangular | |
Food Safe | |
600 x 400 x 420 mm | |
400 mm |