Learn More
Bohlender™ Distributore di flaconi HPLC in polipropilene
Click to view available options
Diametro (sistema metrico) esterno:
4 mm
6 mm
Dimensione della confezione:
Pezzo
Descrizione
- Green glass-fiber reinforced PP screw cap for bottle thread GL 45
- Polypropylene body
- Autoclavable
- Very good chemical resistance
- Available either with four ports with screw joints suitable for tubing O.D. 1.6 to 2.0mm and 2.1 to 4.0mm or with four ports with screw joints suitable for tubing O.D. 6.0 mm
Specifica
Specifica
| Colore chiusura | Green |
| Diametro (sistema metrico) esterno | 6 mm |
| Materiale | Polypropylene |
| Setto | 1Each |
| Materiale chiusura | Glass Fiber |
| Quantità | 1 Ea. |