Learn More
VITLAB™ Becher di misurazione in polipropilene
Click to view available options
Autoclavabile:
Autoclavable
Sì
Capacità (sistema metrico):
50 mL
100 ml
250 ml
Scale:
Raised Scale
Scala in rilievo
Scala stampata in nero su entrambi i lati
Dimensione della confezione:
pezzi 12
pezzi 24
Descrizione
These beakers feature a convenient spout and large handle to make measuring, pouring and disposing of solutions easy and mess-free. Lightweight and transparent plastic beakers are ideal for a variety of lab purposes.
- Stable, easy-grip handle and pour spout for secure handling and mess-free dispensing
- Constructed of durable, lightweight polypropylene that is heat resistant and chemically inert
- Highly transparent with a graduated scale for easy measuring and identification
- Raised scale can’t fade or wipe off and is recommended for autoclaving (at 121°C, 2 bar)
- Printed scale provides maximum contrast for easy measuring (to preserve markings, clean at no higher than 60°C)
- Ideal for a variety of lab purposes, including:
- Preparing solutions and decanting supernatant fluids
- Holding and measuring waste fluids prior to disposal
- Measuring solutions for simple chemical reactions
Specifica
Specifica
| Colore | Trasparente |
| Diametro (sistema metrico) esterno | 50 mm |
| Capacità (sistema metrico) | 100 ml |
| Materiale | PTFE (polipropilene) |
| Da utilizzare con (applicazione) | Per la misurazione del volume |
| Altezza (sistema metrico) | 80 mm |
| Incrementi | 2 mL |
| Quantità | 24 Pk. |
| Tipo | Becher dosatore |
| Autoclavabile | Sì |
| Vedi altri risultati |