Learn More
BRAND™ Polypropylene Thread Adapters
Shop All Brand Products
missing translation for 'orderingAttributeHoverText'
Da utilizzare con (apparecchiatura):
For bottle thread GL 22
For bottle thread GL 24-25
For bottle thread GL 30
For bottle thread S 40
For bottle thread S 54
For bottle thread S 60
For ground joint NS 24/29
missing translation for 'unitSize'
Pezzo
Description
- Thread adapter, Polypropylene (PP), outer thread GL 45, for bottle thread S 40
- Thread Adapters for BRAND™ Bottle Top Dispensers, Polypropylene, 45/40 mm
- For use with Bottle Thread 40 S, Dispensette, Seripettor, Titrette and QuikSip Bottletop Diluters and Dispensers
- The Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use
- Ideal for use with Dispensetter bottletop dispensers
- These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND™ bottletop dispensers and burettes
- These thread adapters help to maximize the compatibility of bottletop dispensers with most reagent bottles
Specifications
Specifications
| Product Type | Adattatore filettato |
| Materiale | Polipropilene |
| Da utilizzare con (apparecchiatura) | For ground joint NS 24/29 |
| Quantità | Confezione da 5 |
| Dimensioni filo | 32 GL |