Learn More
Corning™ Polystyrene Microplates
Sfoglia Tutto Corning Prodotti
Click to view available options
Total Volume (Metric) Well:
0.0128 mL
190 μL
360 μL
Trattamento superficiale:
Non trattata
Sulfhydryl
Dimensione della confezione:
pezzi 100
pezzi 50
Descrizione
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.
- Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
- Recommended working volume of 50μL to 125μL
- Designed for in vitro diagnostic assays
- Untreated surface is hydrophobic in nature
- Color coded strips
- Bar coding available on request
Specifica
Specifica
| Formato | Strip Plate |
| Coperchio | Without Lid |
| Matrice | 8 x 12 |
| Colore | Clear |
| Materiale | Polystyrene |
| No. of Wells | 96 |
| Volume (sistema metrico) di lavorazione | 75 to 200 μL |
| Sterilità | Non-sterile |
| Trattamento superficiale | Sulfhydryl |
| Total Volume (Metric) Well | 360 μL |
| Vedi altri risultati |