Learn More
Euro Protection EPI™ Polyurethane Rainwear
Click to view available options
Colore:
Blue
Yellow
Size:
Large
2X-Large
Dimensione della confezione:
Pezzo
Descrizione
- Waterproof - Welded seams
- Breathable, thick PU/PVC fabric
- Windproof
- Fixed drawstring hood concealed within the high collar
- Central zip with storm flap cover
- Raglan sleeves
- Adjustable cuffs with Velcro™ tab
- 2 lover pockets with flaps
- Ventilated back/underarm ventilation eyelets
- Drawstring waist tightening
- Adjustable lower legs with snaps
- Elacticated waistband
Specifica
Specifica
| Colore | Blue |
| Hood Type | Drawstring |
| Materiale | Polyester |
| Size | Large |
| Tipo di chiusura | Zipper with Storm Flap and Snaps |
| Rivestimento | Polyurethane, PVC |
| Tipo di colletto | High collar with concealed drawstring hood |
| N. di tasche | 2 |
| Tipo di giuntura | Welded |
| Spessore | 300 gsm |
| Vedi altri risultati |