Learn More
Cytiva POM/EPDM Packing Connectors
Sfoglia Tutto Cytiva Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
For use with XK 16 columns
For use with XK 26 columns
Dimensione della confezione:
Pezzo
Descrizione
This packaging connector used to connect another column, being used as a packing reservoir, to a XK 16 or XK 26 column to achieve efficient packing. A properly packed media bed is essential for obtaining optimal results in all forms of liquid chromatography, particularly in gel filtration. The packing connector makes it easy to obtain a well-packed bed of gel filtration and other media in XK 16 and XK 26 Columns, and is especially convenient for fast and easy packing of affinity, IEX, and HIC media to short bed heights.
Specifica
Specifica
| Tipo | Packing Connector |
| Temperatura (sistema metrico) max | 60°C |
| Include | 2 x Sealing ring are delivered with the packing connector |
| Materiale | EPDM, POM |
| Da utilizzare con (apparecchiatura) | For use with XK 16 columns |