missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMC Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA506608
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a polypeptide hormone precursor that undergoes extensive, tissue-specific, post-translational processing via cleavage by subtilisin-like enzymes known as prohormone convertases. There are eight potential cleavage sites within the polypeptide precursor and, depending on tissue type and the available convertases, processing may yield as many as ten biologically active peptides involved in diverse cellular functions. The encoded protein is synthesized mainly in corticotroph cells of the anterior pituitary where four cleavage sites are used; adrenocorticotrophin, essential for normal steroidogenesis and the maintece of normal adrenal weight, and lipotropin beta are the major end products. In other tissues, including the hypothalamus, placenta, and epithelium, all cleavage sites may be used, giving rise to peptides with roles in pain and energy homeostasis, melanocyte stimulation, and immune modulation. These include several distinct melanotropins, lipotropins, and endorphins that are contained within the adrenocorticotrophin and beta-lipotropin peptides. Mutations in this gene have been associated with early onset obesity, adrenal insufficiency, and red hair pigmentation. Alternatively spliced transcript variants encoding the same protein have been described.Specifica
| POMC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01189 | |
| POMC | |
| Full length human recombit protein of human POMC produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1A8 | |
| Unconjugated | |
| POMC | |
| ACTH, CLIP, LPH, MSH, NPP, POC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5443 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto