Learn More
POMC Mouse anti-Human, CoraLite 594, Clone: 3C12C7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL594-66358-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The POMC antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human POMC. This antibody recognizes human, rat, mouse antigen. The POMC antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifica
| POMC | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| 3C12C7 | |
| CoraLite 594 | |
| 100 μL | |
| 5443 | |
| IgG1 |