Learn More
R&D Systems™ Porcine C-Reactive Protein/CRP DuoSet ELISA
Codice prodotto. 30021672
Click to view available options
Quantità:
15 x 96-well Plates
Dimensione della confezione:
1 set
Codice prodotto. 30021672
Marca: R&D Systems™ DY2648
Porcine C-Reactive Protein/CRP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Porcine C-Reactive Protein/CRP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies porcine C-Reactive Protein/CRP.
TRUSTED_SUSTAINABILITY
Specifications
Quantità 15 x 96-well Plates
Coniugato Biotin
Intervallo saggio 78.10 to 5000 pg/mL
Da utilizzare con (applicazione) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.