Learn More
Potassium Hydrogen Phthalate Solution, EU Pharmacopoeia Grade, Fisher Chemical™
Codice prodotto. 12993574 Sfoglia Tutto Fisher Chemical Prodotti
Click to view available options
Quantità:
1000 mL
Dimensione della confezione:
1000mL
Codice prodotto. 12993574
Marca: Fisher Chemical VL 262 1000
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifica
Quantità 1000 mL