Learn More
Potassium Iodide, 10% Solution, Chem-Lab
Product Code. 11945049
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11945049 1 L 1 L
11908454 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11945049 Supplier Chem-Lab Supplier No. CL02.1126.1000
  • ONLINE EXCLUSIVE PRICE
    119.00€
    38.90€ / 1L
    Risparmia 80.10€
    67% Sconto
 Restricted Item
Spedizione stimata: 10-12-2026
Used in iodine isolation, in redox titrations and as a source of iodide in organic synthesis. Potassium Iodide, 10% Solution, Chem-Lab is a solution of 100g KI per liter used in laboratory applications (TMAR).
Potassium iodide is an ionic compound that has been used as a medication (particularly in response to exposure to radioactive iodine). It is a mild reducing agent that is easily oxidized. It is used in iodine isolation, in redox titrations and as a source of iodide in organic synthesis. It may also act as a nucleophilic catalyst for the alkylation of alkyl chlorides, bromides or mesylates.
Chemical Identifiers
CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
Specifications
Quantity 1 L
Formula Weight 166.01g/mol
Concentration or Composition (by Analyte or Components) 10%
Density 1.07g/mL
compliance-icons
Product Identifier
  • Potassium iodide 10% solution
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Potassium iodide