Learn More
Chemsplash PP Bouffant Mop Cap
GREENER_CHOICE
Codice prodotto. p-200105892
Click to view available options
Colore:
Blue
White
Dimensione della confezione:
pezzi 1000
Codice prodotto. 30058199
Marca: Chemsplash 5779.300.999
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
This product is suitable for use in BRC approved work areas and for the Pharmaceutical Industry.
Hand Made Polypropylene Mob Cap
  • Material: 12GSM Polypropylene
  • Hand Made
  • Size: 21 in.
  • With Elastic Band
  • Pack of 100
  • PFAS Free
Description: This product is suitable for use in BRC approved work areas and for the Pharmaceutical Industry. The hand-made mob cap is made from 12GSM polypropylene material and features an elastic band for a secure fit. It is PFAS-free and comes in a pack of 100, making it ideal for both food and pharmaceutical applications.
Specifica
Product Type Bouffant
Size One Size
Materiale Polypropylene
Colore Blue
Monouso Single-use
Senza lattice Yes