missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PPIL2 Recombinant Protein

Marca:  Abnova™ H00023759-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127672

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifica

Specifica

AAH00022
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
83.71
8
Glutathione Sepharose 4 Fast Flow
25 ug
MGKRQHQKDKMYITCAEYTHFYGGKKPDLPQTNFRRLPFDHCSLSLQPFVYPVCTPDGIVFDLLNIVPWLKKYGTNPSNGEKLDGRSLIKLNFSKNSEGKYHCPVLFTVFTNNTHIVAVRTTGNVYAYEAVEQLNIKAKNFRDLLTDEPFSRQDIITLQDPTNLDKFNVSNFYHVKNNMKIIDPDEEKAKQDPSYYLKNTNAETRETLQELYKEFKGDEILAATMKAPEKKKVDKLNAAHYSTGKVSASFTSTAMVPETTHEAAAIDEDVLRYQFVKKKGYVRLHTNKGDLNLELHCDLTPKTCENFIRLCKKHYYDGTIFHRSIRNFVIQGGDPTGTGTGGESYWGKPFKDEFRPNLSHTGRGILSMANSGPNSNRSQFFITFRSCAYLDKKHTIFGRVVGGFDVLTAMENVESDPKTDRPKEEIRIDATTVFVDPYEEADAQIAQERKTQLKVAPETKVKSSQPQAGSQGPQTFRQGVGKYINPAATEQQRKSPQPVPLSPCPRRSPVGVLGTSAPGSSRLPDDH
CYC4/CYP60/Cyp-60/FLJ39930/MGC33174/MGC787/hCyP-60
PPIL2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
23759
PPIL2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPIL2
Human
Recombinant
Solution
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali