missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP3CB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA570325
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.
Specifica
| PPP3CB | |
| Polyclonal | |
| Unconjugated | |
| PPP3CB | |
| 1110063J16Rik; calcineurin A; calcineurin A beta; Calcineurin A protein; calcineurin A2; Calcineurin subunit A beta; calmodulin-dependent calcineurin A subunit beta isoform; CALNA2; Calnb; CAM-PRP catalytic subunit; CaN Abeta; CNA beta; CNA2; Cnab; CnAbeta; hypothetical protein LOC550554; PP2BA beta; PP2Bbeta; Ppp3cb; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3 catalytic subunit beta; protein phosphatase 3, catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; protein phosphatase-2Bb; RP11-345K20.1; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit alpha; wu:fq19c08; zgc:110141 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24675 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P20651 | |
| PPP3CB | |
| synthetic peptide directed towards the middle region of rat PPP3CB. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto