Learn More
Invitrogen™ PPP3CB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA596249
Descrizione
Immunogen sequence: IIRAHEAQDA GYRMYRKSQT TGFPSLITIF SAPNYLDVYN NKAAVLKYEN NVMNIRQFNC SPHPYWLPNF MDVFTWSLPF VGEKVTEMLV NVLSICSDDE LMTEGEDQFD VGSAAARKEI IRNKIRAIGK MARVFSVLRE ESESVLTLKG LTPTGMLPSG VLAGGRQTLQ SATVEAIEAE KAIRGFSPPH RICSFEEAKG LDRINERMPP RKDAVQQDGF NSLNTAHATE NHGTGNHTAQ; Positive Samples: Mouse brain.
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.
Specifica
| PPP3CB | |
| Polyclonal | |
| Unconjugated | |
| PPP3CB | |
| 1110063J16Rik; calcineurin A; calcineurin A beta; Calcineurin A protein; calcineurin A2; Calcineurin subunit A beta; calmodulin-dependent calcineurin A subunit beta isoform; CALNA2; Calnb; CAM-PRP catalytic subunit; CaN Abeta; CNA beta; CNA2; Cnab; CnAbeta; hypothetical protein LOC550554; PP2BA beta; PP2Bbeta; Ppp3cb; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3 catalytic subunit beta; protein phosphatase 3, catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; protein phosphatase-2Bb; RP11-345K20.1; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit alpha; wu:fq19c08; zgc:110141 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19056, 5532 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.99 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16298, P48453 | |
| PPP3CB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 286-525 of human PPP3CB (NP_0011358251). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.