Learn More
OHAUS™ Precision Balance - PR Series
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Modello:
PR1602M
PR2202M
PR223M
PR4201M
PR4202M
PR423M
PR523M
PR6201M
Dimensione della confezione:
Pezzo
Descrizione
- The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
- Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
- The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
Specifica
Specifica
| Tipo di collegamento | RS232 (one port included) |
| Descrizione | User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator |
| Capacità | 220.00 g |
| Tipo di display | Backlit LCD |
| Leggibilità | 0.0010 g |
| Da utilizzare con (applicazione) | Basic Weighing, Parts Counting, Percent Weighing |
| Linearità | 0.0020 g |
| Modello | PR223M |
| Ripetibilità | 0.0010 g |
| Tempo di stabilizzazione | 2.000 s |
| Vedi altri risultati |