Learn More
Uvex Preformed Mask silv-Air p 7220 FFP2
Codice prodotto. 17502955 Sfoglia Tutto Uvex Prodotti
Click to view available options
:
Filter Type: FFP2
Codice prodotto. 17502955
Marca: Uvex 8707220
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering preformed mask with exhalation valve to reduce exhalation resistance and heat and moisture build-up inside the mask
  • Metal-free processing
  • Additional activated carbon filter against odors, gases and vapors below threshold values
  • Seamless headband for a comfortable, secure fit
  • Color: silver, anthracite
Protection Features
  • FFP2 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Seamless headband for a comfortable, secure fit
  • Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
TRUSTED_SUSTAINABILITY
Specifica
Certifications/Compliance (Europe) EN 149 (D marked)
Monouso Single-use
Materiale Polypropylene, Polyester
Product Type Preformed with Carbon Filter
Size Standard
With Exhalation Valve Sì
Efficienza filtro FFP2