missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Pressinoic Acid

Codice prodotto. 16489558
Encompass_Preferred
Click to view available options
Quantità:
10 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16489558

Marca: MedChemExpress HYP1487

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Pressinoic Acid is a synthetic hexapeptide with potent corticotrophin-releasing activity. Pressinoic Acid is also an oxytocin inhibitor; it induces maternal behavior.

Specifica

CAS 35748-51-7
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Da utilizzare con (applicazione) Neuroscience-Neuromodulation
Peso molecolare 774.86 g/mol
Product Type Peptides
Quantità 10 mg
Shipping Condition Room temperature
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.