Learn More
JULABO PRESTO Process Systems
GREENER_CHOICE
Codice prodotto. p-7227811
Click to view available options
Modello:
A38
A41
A70
W41
Dimensione della confezione:
Pezzo
Codice prodotto. 17714869
Marca: JULABO 9420381.N1
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The PRESTO systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
  • Wide working temperature ranges without changing fluids
  • Ethernet interface
  • SD-Card slot
  • pace optimized design creates more room directly next to the units
  • Connections for USB, Ethernet, RS232, and Alarm output
Specifica
Temperature Range (Metric) -30°C to +250°C
Dimensioni (L x P x A) 29.5 x 13 x 26.4 in.
Modello A38
Tipo Process System