missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRMT5 Monoclonal Antibody (PRMT5-21)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA125470
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Recommended positive controls: G361.
PRMT5 (protein arginine N-methyltransferase 5) is an arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. PRMT5 specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins Sm D1 and Sm D3; such methylation is required for the assembly and biogenesis of snRNP core particles.
Specifica
| PRMT5 | |
| Monoclonal | |
| 2.4 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| A7YW45, O14744, Q8CIG8 | |
| PRMT5 | |
| GST-PRMT5 containing the C-terminal region of the human PRMT5 (amino acids 315-637). | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Chicken | |
| Antibody | |
| IgG2a |
| ChIP Assay, ELISA, Western Blot | |
| PRMT5-21 | |
| Unconjugated | |
| PRMT5 | |
| 72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs | |
| Mouse | |
| Purified | |
| RUO | |
| 100768836, 101750306, 10419, 27374, 364382, 480242, 515594 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto