missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pro-Detect™ Rapid Antibody Isotyping Assay Kit, rat
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
10 tests
Dimensione della confezione:
pezzi 10
Descrizione
Includes
10 assays
- Uses membrane-based strips coated with dye-bound conjugated antibodies that provide a visual, color readout of the monoclonal antibody isotype within ten minutes.
- Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
- Simple, fast method—dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
- Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentration
- Specific—assay does not cross-react with common lysis and protein preparation reagents
- Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
- Long shelf life—stable for at least one year at 4°C
Specifica
Specifica
| Content And Storage | • 10 Rapid Antibody Isotyping Assay Strips 1 • 10 Rapid Antibody Isotyping Assay Strips 2 • Sample Diluent, 4 mL Sufficient for 10 isotype determinations Store at 2–8°C. |
| Analiti rilevabili | IgG1, IgG2a, IgG2b; IgG2c |
| Saggio | Isotyping Assay |
| Metodo di rilevamento | Chromogenic |
| Linea di prodotti | Pro-Detect |
| Product Type | Assay |
| Specificità | Antibody |
| Sufficiente per | 10 tests |
| Target Species | Rat |
| Da utilizzare con (applicazione) | Lateral Flow Rapid Assay |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto