missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pro-Detect™ Rapid MBP Assay Kit
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
10 Assays
Dimensione della confezione:
pezzi 10
Descrizione
Includes
10 assays
- Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a visual, color readout for detection within ten minutes.
- Detect proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.
- Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
- Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
- Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
- Specific—assay does not cross-react with common lysis and protein preparation reagents
- Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
- Long shelf life—stable for at least one year at 4°C
Specifica
Specifica
| Content And Storage | • 10 Rapid Assay Strips • Sample Diluent, 15 mL Sufficient for 10 assay determinations Store at 2–8°C. |
| Analiti rilevabili | MBP-tagged proteins |
| Saggio | Lateral Flow Rapid Assay |
| Metodo di rilevamento | Chromogenic |
| Linea di prodotti | Pro-Detect |
| Product Type | Assay |
| Specificità | Protein tag |
| Sufficiente per | 10 Assays |
| Da utilizzare con (applicazione) | Lateral Flow Rapid Assay |
| Quantità | 10 Assays |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto