Learn More
Corning™ ProCulture™ Fittings
Sfoglia Tutto Corning Prodotti
Click to view available options
Product Type:
Combination Sidearm
Dual Sidearm
Media Handling Dual Sidearm
Sidearm
Triple Sidearm
Descrizione:
1 Tube
2 Tubes, Angled
2 Tubes, Straight
3 Tubes, Angled
Diameter (Metric) Tubing:
18 mm
3.17 mm
6.35 mm
6.35/3.17 mm
Da utilizzare con (apparecchiatura):
1 L Sidearm Spinner Flask
15 L Sidearm Spinner Flask
3 L Sidearm Spinner Flask
500 L Sidearm Spinner Flask
6 L Sidearm Spinner Flask
8 L Sidearm Spinner Flask
8, 15 L Sidearm Spinner Flask
Large Spinner Flasks w/ Vertical Sidearms
Dimensione della confezione:
Pezzo
Descrizione
- Used to aseptically transfer medium, sparge the cell culture medium directly or add gases to the head space above the cell culture medium
- Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
- One or two 316 stainless steel tubes extend to the bottom of the flask; the other is a shorter 6 length.
- Both tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
- The fittings are completely autoclavable
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
Specifica
Specifica
| Descrizione | 2 Tubes, Angled |
| Diameter (Metric) Tubing | 6.35 mm |
| Materiale | Stainless Steel |
| Da utilizzare con (applicazione) | Used to sparge the cell culture medium with gases to enhance cell production. |
| Da utilizzare con (apparecchiatura) | 15 L Sidearm Spinner Flask |
| Include | Insert, O-ring, sealing cap |
| N. per custodia | Each |
| Product Type | Sidearm |
| Autoclavabile | Autoclavable |
| Sterilità | Non-sterile |